Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zdhhc18
Entrez Gene ID
100771709
Gene Name
zinc finger, DHHC-type containing 18
Gene Aliases
I79_016193
Chromosome Location
Chr.Un|NW_003614517: 344096 - 377425 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648923.1 XP_007647113.1 7 - 8 857 83
XM_007648924.1 XP_007647114.1 7 - 8 857 83
XM_007648925.1 XP_007647115.1 7 - 8 857 83
XM_007648926.1 XP_007647116.1 7 - 8 857 83
XM_007648927.1 XP_007647117.1 7 - 8 876 83

View Full Product Details