Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1rl
Entrez Gene ID
100760066
Gene Name
complement component 1, r subcomponent-like
Gene Aliases
I79_001434
Chromosome Location
Chr.Un|NW_003613610: 4103389 - 4122445 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496073.2 XP_003496121.1 3 - 4 557 66

View Full Product Details