Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C1rl
Entrez Gene ID
100760066
Gene Name
complement component 1, r subcomponent-like
Gene Aliases
I79_001434
Chromosome Location
Chr.Un|NW_003613610: 4103389 - 4122445 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003496073.2 XP_003496121.1 4 - 5 668 69
GenBank mRNA JI873923.1 - 1 - 2 70 69
JP059183.1 - 1 - 2 70 69

View Full Product Details