Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cc2d2b
Entrez Gene ID
100763170
Gene Name
coiled-coil and C2 domain containing 2B
Gene Aliases
I79_003389
Chromosome Location
Chr.Un|NW_003613664: 160262 - 205696 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497772.1 XP_003497820.1 9 - 10 1079 86

View Full Product Details