Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mesp2
Entrez Gene ID
100751921
Gene Name
mesoderm posterior 2 homolog (mouse)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617731: 27315 - 29089 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007607523.1 XP_007605713.1 1 - 2 183 89

View Full Product Details