Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec11a
Entrez Gene ID
100755020
Gene Name
C-type lectin domain family 11, member A
Gene Aliases
I79_018464
Chromosome Location
Chr.Un|NW_003614844: 253594 - 255721 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510199.2 XP_003510247.1 1 - 2 144 101

View Full Product Details