Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam69b
Entrez Gene ID
100754761
Gene Name
family with sequence similarity 69, member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617120: 50108 - 57891 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514922.2 XP_003514970.1 1 - 2 162 81

View Full Product Details