Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Xk
Entrez Gene ID
100762544
Gene Name
X-linked Kx blood group (McLeod syndrome)
Gene Aliases
I79_016005
Chromosome Location
Chr.Un|NW_003614493: 373578 - 411232 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508158.2 XP_003508206.2 3 - 4 402 64

View Full Product Details