Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp41
Entrez Gene ID
100757735
Gene Name
ZFP41 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614880: 291172 - 302059 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510439.2 XP_003510487.1 2 - 3 1030 75

View Full Product Details