Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rbm44
Entrez Gene ID
100760953
Gene Name
RNA binding motif protein 44
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616745: 3760 - 21244 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007654665.1 XP_007652855.1 8 - 9 2202 102

View Full Product Details