Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr65
Entrez Gene ID
100769099
Gene Name
WD repeat domain 65
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613906: 823880 - 895262 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502655.2 XP_003502703.1 21 - 22 3558 94
XM_007643294.1 XP_007641484.1 21 - 22 3363 94

View Full Product Details