Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cplx4
Entrez Gene ID
100755686
Gene Name
complexin 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614319: 22661 - 36039 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507024.1 XP_003507072.1 1 - 2 169 81

View Full Product Details