Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prap1
Entrez Gene ID
100772682
Gene Name
proline-rich acidic protein 1
Gene Aliases
I79_020661
Chromosome Location
Chr.Un|NW_003615308: 274359 - 289919 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512005.1 XP_003512053.1 2 - 3 184 65

View Full Product Details