Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Stxbp3
Entrez Gene ID
100757848
Gene Name
syntaxin binding protein 3
Gene Aliases
Stxbp3a
Chromosome Location
Chr.Un|NW_003617055: 22396 - 64662 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514867.2 XP_003514915.1 9 - 10 911 81
XM_007654905.1 XP_007653095.1 9 - 10 912 81
GenBank mRNA JI894043.1 - 9 - 10 904 81
JP062973.1 - 9 - 10 904 81

View Full Product Details