Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pxmp2
Entrez Gene ID
100774960
Gene Name
peroxisomal membrane protein 2, 22kDa
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613859: 1585599 - 1596621 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501730.2 XP_003501778.1 3 - 4 395 82

View Full Product Details