Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klk4
Entrez Gene ID
100757055
Gene Name
kallikrein-related peptidase 4
Gene Aliases
I79_018451
Chromosome Location
Chr.Un|NW_003614844: 127468 - 131268 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510206.1 XP_003510254.1 4 - 5 624 67

View Full Product Details