Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asphd2
Entrez Gene ID
100773627
Gene Name
aspartate beta-hydroxylase domain containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614271: 510822 - 516300 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647213.1 XP_007645403.1 2 - 3 943 56

View Full Product Details