Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asb10
Entrez Gene ID
100761357
Gene Name
ankyrin repeat and SOCS box containing 10
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613842: 959524 - 968863 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501294.1 XP_003501342.1 3 - 4 1058 90

View Full Product Details