Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem50a
Entrez Gene ID
100752566
Gene Name
transmembrane protein 50A
Gene Aliases
I79_022302
Chromosome Location
Chr.Un|NW_003615875: 6837 - 17181 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003513351.2 XP_003513399.1 5 - 6 481 114
GenBank mRNA JP054972.1 - 1 - 1 503 114

View Full Product Details