Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Il36b
Entrez Gene ID
100773970
Gene Name
interleukin 36, beta
Gene Aliases
I79_025379
Chromosome Location
Chr.Un|NW_003619099: 8320 - 23817 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515696.1 XP_003515744.1 2 - 3 209 100

View Full Product Details