Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cmtm8
Entrez Gene ID
100768898
Gene Name
CKLF-like MARVEL transmembrane domain containing 8
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613697: 2663510 - 2720655 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007630299.1 XP_007628489.1 3 - 4 500 76

View Full Product Details