Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msmb
Entrez Gene ID
100756196
Gene Name
microseminoprotein, beta-
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613589: 1626347 - 1647669 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640069.1 XP_007638259.1 1 - 2 67 87

View Full Product Details