Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasd2
Entrez Gene ID
100766916
Gene Name
RASD family, member 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614771: 12754 - 16455 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509810.2 XP_003509858.1 1 - 2 275 120

View Full Product Details