Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txnl4a
Entrez Gene ID
100760868
Gene Name
thioredoxin-like 4A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613863: 463664 - 478504 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501837.2 XP_003501885.1 1 - 2 154 107

View Full Product Details