Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Parp15
Entrez Gene ID
100770244
Gene Name
poly (ADP-ribose) polymerase family, member 15
Gene Aliases
I79_007463
Chromosome Location
Chr.Un|NW_003613831: 1923846 - 1936010 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501112.1 XP_003501160.1 10 - 11 1312 78

View Full Product Details