Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100757639
Entrez Gene ID
100757639
Gene Name
putative olfactory receptor GPCRLTM7
Gene Aliases
I79_019670
Chromosome Location
Chr.Un|NW_003615056: 99040 - 99987 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651742.1 XP_007649932.1 1 - 1 524 97

View Full Product Details