Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nnat
Entrez Gene ID
100757948
Gene Name
neuronatin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613641: 1384046 - 1388646 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497102.2 XP_003497150.1 1 - 2 109 101

View Full Product Details