Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Scgb1d4
Entrez Gene ID
100757048
Gene Name
secretoglobin, family 1D, member 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614577: 535800 - 540737 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508752.1 XP_003508800.1 1 - 2 52 110

View Full Product Details