Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
Cdr1
Entrez Gene ID
100769322
Gene Name
cerebellar degeneration-related protein 1, 34kDa
Gene Aliases
I79_019853
Chromosome Location
Chr.Un|NW_003615089: 50947 - 52947 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651876.1 XP_007650066.1 - 1624 105

View Full Product Details