Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyz
Entrez Gene ID
100770686
Gene Name
lysozyme
Gene Aliases
I79_002688
Chromosome Location
Chr.Un|NW_003613642: 3332447 - 3337902 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497141.2 XP_003497189.1 2 - 2 220 84

View Full Product Details