Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pcdh20
Entrez Gene ID
100757789
Gene Name
protocadherin 20
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613979: 360290 - 364212 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503541.2 XP_003503589.1 2 - 2 2718 93

View Full Product Details