Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rgag4
Entrez Gene ID
100758014
Gene Name
retrotransposon gag domain containing 4
Gene Aliases
I79_016850
Chromosome Location
Chr.Un|NW_003614593: 285981 - 288827 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508819.2 XP_003508867.2 2 - 2 1126 108

View Full Product Details