Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Znhit2
Entrez Gene ID
100766069
Gene Name
zinc finger, HIT-type containing 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003616756: 25900 - 27728 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514589.2 XP_003514637.1 1 - 1 1226 73

View Full Product Details