Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rwdd2b
Entrez Gene ID
100768876
Gene Name
RWD domain containing 2B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613595: 500108 - 511516 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495508.2 XP_003495556.1 5 - 5 2618 107
XM_007641765.1 XP_007639955.1 5 - 5 2556 107

View Full Product Details