Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Crygs
Entrez Gene ID
100765744
Gene Name
crystallin, gamma S
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614504: 590322 - 595728 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508222.2 XP_003508270.1 3 - 3 430 105

View Full Product Details