Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Utf1
Entrez Gene ID
100770383
Gene Name
undifferentiated embryonic cell transcription factor 1
Gene Aliases
I79_020651
Chromosome Location
Chr.Un|NW_003615308: 150978 - 152027 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652512.1 XP_007650702.1 1 - 1 223 68

View Full Product Details