Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WBP1
Entrez Gene ID
102173394
Gene Name
WW domain binding protein 1
Gene Aliases
-
Chromosome Location
Chr.11: 10012564 - 10015003 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686330.3 XP_005686387.1 2 - 3 332 71

View Full Product Details