Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNB1
Entrez Gene ID
102186994
Gene Name
cholinergic receptor nicotinic beta 1 subunit
Gene Aliases
-
Chromosome Location
Chr.19: 26933757 - 26944480 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693503.2 XP_005693560.2 8 - 9 1106 64

View Full Product Details