Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTG2
Entrez Gene ID
102178279
Gene Name
actin gamma 2, smooth muscle
Gene Aliases
-
Chromosome Location
Chr.11: 10599119 - 10623482 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686352.3 XP_005686409.1 6 - 7 902 82
XM_005686353.3 XP_005686410.1 5 - 6 773 82

View Full Product Details