Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UFC1
Entrez Gene ID
102169436
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
-
Chromosome Location
Chr.3: 111780390 - 111784314 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046411.1 XP_017901900.1 1 - 2 394 74

View Full Product Details