The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285602.1 | NP_001272531.1 | 2 - 3 | 654 | 84 |
GenBank mRNA | HM204707.1 | - | 2 - 3 | 654 | 84 |
JT651870.1 | - | 3 - 4 | 661 | 84 |