Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR2
Entrez Gene ID
102180272
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 2
Gene Aliases
-
Chromosome Location
Chr.3: 105408575 - 105411678 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677361.3 XP_005677418.1 2 - 3 388 98
GenBank mRNA KA349045.1 - 2 - 3 317 98

View Full Product Details