Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TRAPPC4
- Entrez Gene ID
-
108633163
- Gene Name
- trafficking protein particle complex 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 52754517 - 52758158 on Build ARS1
- Gene Symbol
- RPS25
- Entrez Gene ID
-
102171627
- Gene Name
- ribosomal protein S25
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 52758217 - 52760175 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KA331002.1 |
- |
2 - 3 |
475 |
59 |
View Full Product Details