The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_018060747.1 | XP_017916236.1 | 2 - 3 | 459 | 79 |
XM_018060748.1 | XP_017916237.1 | 3 - 4 | 522 | 79 |