Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C13H20orf27
Entrez Gene ID
102180506
Gene Name
chromosome 13 C20orf27 homolog
Gene Aliases
-
Chromosome Location
Chr.13: 50785602 - 50799452 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057399.1 XP_017912888.1 4 - 5 1280 83
GenBank mRNA JO333774.2 - 4 - 5 495 83
JO413267.2 - 2 - 3 768 83

View Full Product Details