Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPL19
Entrez Gene ID
102170432
Gene Name
mitochondrial ribosomal protein L19
Gene Aliases
-
Chromosome Location
Chr.11: 43620249 - 43631032 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686638.3 XP_005686695.2 2 - 3 251 97
GenBank mRNA JT680139.1 - 6 - 7 1207 97

View Full Product Details