Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL1
Entrez Gene ID
102184927
Gene Name
angiopoietin like 1
Gene Aliases
-
Chromosome Location
Chr.16: 58825677 - 58849018 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690918.3 XP_005690975.1 3 - 4 1509 85

View Full Product Details