Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNS
Entrez Gene ID
100750233
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
Chromosome Location
Chr.5: 48392750 - 48439234 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285690.1 NP_001272619.1 6 - 7 834 67
GenBank mRNA JO373133.2 - 6 - 7 846 67
U17694.1 - 6 - 7 834 67

View Full Product Details