Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPRC5B
Entrez Gene ID
102178968
Gene Name
G protein-coupled receptor class C group 5 member B
Gene Aliases
-
Chromosome Location
Chr.25: 17314610 - 17335943 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040564.1 XP_017896053.1 2 - 3 1150 77
GenBank mRNA JO434014.2 - 2 - 3 1153 77

View Full Product Details