Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CFAP410
Entrez Gene ID
102188261
Gene Name
cilia and flagella associated protein 410
Gene Aliases
C1H21orf2
Chromosome Location
Chr.1: 144256566 - 144267295 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051920.1 XP_017907409.1 2 - 3 320 83
XM_018051928.1 XP_017907417.1 2 - 3 321 83
XM_018051937.1 XP_017907426.1 2 - 3 321 83

View Full Product Details